missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human CD28 (soluble) Fc Recombinant Protein, PeproTech®

Code produit. 17842973
Click to view available options
Quantité:
1 mg
100 μg
20 μg
250 μg
500 μg
Conditionnement:
100µg
1mg
20µg
250µg
500µg
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Code produit. 17842973

Marque: Gibco™ 31034500UG

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Recombinant Protein

PeproTech's CHO cell-derived Recombinant Human sCD28 Fc is a glycosylated, disulfide-linked homodimer of 734 amino acid residues whose monomer consists of the 134-amino-acid length extracellular portion of CD28 fused to the 231-amino-acid length Fc portion of human IgG1 by two glycines. The calculated molecular weight of Recombinant Human sCD28 Fc dimer is 82.4 kDa; however, due to glycosylation, the monomer and dimer migrate at apparent molecular weights of approximately 60-70 kDa and 150-160 kDa by SDS-PAGE analysis under reducing conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

CD28 antigen is a 44 kDa disulfide linked homodimeric T cell specific surface glycoprotein. CD28 is a cell adhesion molecule of the immunoglobulin superfamily which is constitutively expressed on most peripheral blood T lymphocytes. Moreover, CD28 is the critical T cell costimulatory receptor that provides the cell the important second activation signal by binding CD80 and CD86 which are expressed by antigen presenting cells. In addition to its co-stimulation role, CD28 functions by preventing T cells from entering an anergic-hyporesponsive state or from undergoing premature apoptotic cell death. In murine peripheral lymphoid organs and in the blood, all CD4+ and CD8+ T cells express CD28. In the thymus, CD28 expression is highest on immature CD3-, CD8+ and CD4+8+ cells, and on CD4-8- cells that express alpha/beta and gamma/delta TCR. The level of CD28 on mature CD4+ and CD8+ alpha/beta TCR+ thymocytes is two- to fourfold lower than on the immature cells. Diseases associated with CD28 dysfunction include mycosis fungiodes and Sezary's Disease.
TRUSTED_SUSTAINABILITY

Spécification

Numéro d’adhésion P10747
À utiliser avec (application) ELISA, Functional Assay
Formule protein with no preservative
Identification génétique (Entrez) 940
Poids moléculaire 82.4 kDa
Nom Human CD28 (soluble) Fc
Quantité 500 μg
Source CHO cells
État réglementaire RUO
Concentration en endotoxines <1 EU/ μg
Alias de gène antigen CD28; CD28; CD28 antigen; CD28 antigen (Tp44); CD28 isoform; CD28 isoform 2; Cd28 molecule; CD28 precursor protein; CD28 protein; CD28 protein precursor; CD28RNA; cell surface protein; CHT28; contains partial extracellular domain; costimulatory molecule B7 receptor CD28; EGK_04709; MGC138290; sCD28; soluble CD28; T44; T-cell costimulatory molecule CD28; T-cell costimulatory molecule Tp44; T-cell-specific surface glycoprotein CD28; T-cell-specific surface glycoprotein CD28 homolog; TP44
Nom usuel CD28
Symbole de gène(s) Cd28
Activité biologique Determined by its ability to bind its principal binding partner (B7-1) in a functional ELISA.
Conjugué Unconjugated
Recombinant Recombinant
Marqueur de protéine Fc-tag
Séquence NKILVKQSPM LVAYDNAVNL SCKYSYNLFS REFRASLHKG LDSAVEVCVV YGNYSQQLQV YSKTGFNCDG KLGNESVTFY LQNLYVNQTD IYFCKIEVMY PPPYLDNEKS NGTIIHVKGK HLCPSPLFPG PSKPGGPKSC DKTHTCPPCP APELLGGPSV FLFPPKPKDT LMISRTPEVT CVVVDVSHED PEVKFNWYVD GVEVHNAKTK PREEQYNSTY RVVSVLTVLH QDWLNGKEYK CKVSNKALPA PIEKTISKAK GQPREPQVYT LPPSRDELTK NQVSLTCLVK GFYPSDIAVE WESNGQPENN YKTTPPVLDS DGSFFLYSKL TVDKSRWQQG NVFSCSVMHE ALHNHYTQKS LSLSPGK
Contenu et stockage -20°C
Système d’expression CHO cells
Forme Lyophilized
Pureté ou qualité ≥ 95% by SDS-PAGE gel and HPLC analyses.
Afficher plus Afficher moins
Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis