missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human CDNF Recombinant Protein, PeproTech®
Click to view available options
Quantité:
1 mg
100 μg
25 μg
250 μg
5 μg
500 μg
Conditionnement:
100µg
1mg
250µg
25µg
500µg
5µg
Description
Recombinant Human CDNF is an 18.5 kDa protein consisting of 162 amino acids, including 8 cysteine residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
The conserved dopamine neurotrophic factor (CDNF) is a neurotrophic factor for dopaminergic neurons and highly homologous to the mesencephalic-astrocyte-derived neuro-trophic factor. Somewhat surprisingly, CDCF is expressed at higher levels in tissues such as heart, skeletal muscle and testes than in brain. Similar to the glial cell line-derived neurotrophic factor (GDNF), CDNF can prevent the 6-hydroxylamine (6 OHDA) induced degeneration of dopaminergic neurons in a rat model of Parkinsons disease. Furthermore, CDNF was able to restore the dopaminergic function and prevent the degeneration of dopaminergic neurons in the substantia nigra, suggesting that CDNF might be suitable for the treatment of Parkinsons disease. At least two isoforms of CDNF are known to exist.
Spécification
Spécification
| Numéro d’adhésion | Q49AH0 |
| À utiliser avec (application) | Functional Assay |
| Formule | protein with no preservative |
| Identification génétique (Entrez) | 441549 |
| Poids moléculaire | 18.5 kDa |
| Nom | Human CDNF |
| Quantité | 25 μg |
| Source | E. coli |
| État réglementaire | RUO |
| Concentration en endotoxines | <1 EU/ μg |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu