missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human CIRH1A (aa 8-89) Control Fragment Recombinant Protein
Click to view available options
Quantité:
100 μl
Conditionnement:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (96%), Rat (96%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-111179 (PA5-111179. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
CIRH1A encodes a WD40-repeat-containing protein that is localized to the nucleolus. Mutation of this gene causes North American Indian childhood cirrhosis, a severe intrahepatic cholestasis that results in transient neonatal jaundice, and progresses to periportal fibrosis and cirrhosis in childhood and adolescence.
Spécification
Spécification
| Numéro d’adhésion | Q969X6 |
| Concentration | ≥5.0 mg/mL |
| À utiliser avec (application) | Blocking Assay, Control |
| Formule | 1 M urea, PBS with no preservative; pH 7.4 |
| Identification génétique (Entrez) | 84916 |
| Nom | Human CIRH1A (aa 8-89) Control Fragment |
| Quantité | 100 μl |
| État réglementaire | RUO |
| Alias de gène | CIRH1A; Cirhin; cirrhosis, autosomal recessive 1 A (cirhin); cirrhosis, autosomal recessive 1 A (human); cPERP-E; Kiaa1988; Naic; Teg-292; testis expressed gene 292; testis-expressed gene 292; testis-expressed gene 292 protein; Te x 292; U3 small nucleolar RNA-associated protein 4 homolog; UTP4; UTP4 small subunit (SSU) processome component; UTP4 small subunit processome component; UTP4, small subunit (SSU) processome component, homolog; UTP4, small subunit processome component |
| Nom usuel | CIRH1A |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu