missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human CXCL1 (GRO alpha) Recombinant Protein
Click to view available options
Quantité:
10 μg
100 μg
25 μg
500 μg
Conditionnement:
100µg
10µg
25µg
500µg
Description
CXCL1, CXCL2, and CXCL3, also known respectively as GRO alpha, GRO beta (MIP-2 alpha) and GRO gamma (MIP-2 beta), are members of the CXC subfamily of chemokines. Mature CXCL1/2/3 proteins bind with high affinity to the IL-8 receptor type B and are potent neutrophil attractants and activators. In vitro, the processed forms GRO-alpha(4-73), GRO-alpha(5-73) and GRO-alpha(6-73) show a 30-fold higher chemotactic activity.
Specifications
Specifications
| Numéro d’adhésion | P09341 |
| Concentration | 200 μg/mL |
| À utiliser avec (application) | Control, ELISA, Western Blot |
| Formule | PBS with no preservative |
| Identification génétique (Entrez) | 2919 |
| Poids moléculaire | 7.8 kD |
| Nom | Human CXCL1 (aa 35-107) |
| Quantité | 10 μg |
| Source | E. coli |
| Conditions de stockage | -20°C |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction