Learn More
Invitrogen™ Human Cyclin O (aa 180-262) Control Fragment Recombinant Protein
Click to view available options
Quantité:
100 μl
Conditionnement:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (87%), Rat (87%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-145141 (PA5-145141. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Spécification
Spécification
| Numéro d’adhésion | P22674 |
| Concentration | ≥5.0 mg/mL |
| À utiliser avec (application) | Blocking Assay, Control |
| Formule | 1 M urea, PBS with no preservative; pH 7.4 |
| Identification génétique (Entrez) | 10309 |
| Nom | Human Cyclin O (aa 180-262) Control Fragment |
| Quantité | 100 μl |
| État réglementaire | RUO |
| Alias de gène | C86987; CCNO; Ccnu; CILD29; cyclin domain containing; cyclin O; cyclin U; cyclin-like uracil-DNA glycosylase; cyclin-O; E130318K11; RGD1565217; UDG2; Ung2; uracil DNA glycosylase 2; uracil-DNA glycosylase 2 |
| Nom usuel | Cyclin O |
| Afficher plus |