missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human DLL4 (soluble) Recombinant Protein, PeproTech®
Click to view available options
Quantité:
1 mg
100 μg
25 μg
250 μg
5 μg
500 μg
Conditionnement:
100µg
1mg
250µg
25µg
500µg
5µg
Description
Recombinant Human sDLL-4 is a 54.3 kDa glycoprotein containing 498 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Delta-like ligand 4 (DLL4), also know as Delta-4, is a homologue of the Drosophila delta protein, and functions as a transmembrane bound ligand to the notch receptor Notch1. DLL4 is expressed in the vasculature and plays a critical role in vascular development. It is induced by vascular endothelial growth factor (VEGF), as a negative feedback regulator to regulate angiogenic sprouting and promote the formulation of a differentiated vascular network. DLL4 has been found to be strongly expressed in tumor vessels of clear-cell renal tumors and bladder cancer, and inhibition of DLL4 results in increased vascular proliferation but defective maturation. This in turn leads to a decrease in tumor growth, with no apparent toxicity.
Spécification
Spécification
| Numéro d’adhésion | Q9NR61 |
| À utiliser avec (application) | ELISA, Functional Assay, Western Blot |
| Formule | protein with no preservative |
| Identification génétique (Entrez) | 54567 |
| Poids moléculaire | 54.3 kDa |
| Nom | Human DLL4 (soluble) |
| Quantité | 250 μg |
| Source | HEK293 cells |
| État réglementaire | RUO |
| Concentration en endotoxines | <1 EU/ μg |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu