missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human E-selectin (CD62E) Recombinant Protein, PeproTech®

Code produit. 17891453
Click to view available options
Quantité:
1 mg
10 μg
100 μg
250 μg
50 μg
500 μg
Conditionnement:
100µg
10µg
1mg
250µg
500µg
50µg
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Code produit. 17891453

Marque: Gibco™ 150151MG

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Recombinant Protein

Recombinant Human E-selectin is a 58.6 kDa protein containing 535 amino acid residues, corresponding to the extracellular portion of the full length protein. Due to glycosylation, E-selectin migrates at an apparent molecular weight of approximately 65-85 kDa by SDS-PAGE analysis under reducing conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Leukocytes adhere to the blood vessel endothelium during extravasation in postcapillary venules of lymph nodes. In addition, leukocyte adhesion occurs in the capillaries and small venules at any site in the body after onset of inflammation. This response is immediate and involves a cascade of adhesion receptors. At the endothelial surface members of the selectin and immunoglobulin superfamilies participate in this cascade. Selectins are C-type cell surface lectins that play a role in leukocyte adhesion to the blood vessel wall endothelium. E-selectin (CD62E) is an endothelial cell specific selectin that is expressed only after activation with proinflammatory cytokines. In vitro experiments have shown that IL-1, TNFalpha and bacterial wall components like lipopolysaccharides induce the transcription of E-selectin in a NFkB dependent signaling cascade. E-selectin has been associated with blood vessel endothelium in diverse inflammatory situations.
TRUSTED_SUSTAINABILITY

Spécification

Numéro d’adhésion P16581
À utiliser avec (application) Functional Assay
Formule protein with no preservative
Identification génétique (Entrez) 6401
Poids moléculaire 58.6 kDa
Nom Human E-selectin (CD62E)
Quantité 1 mg
Source CHO cells
État réglementaire RUO
Concentration en endotoxines <1 EU/ μg
Alias de gène CD62 antigen-like family member E; CD62E; e selectin; Elam; ELAM1; ELAM-1; endothelial adhesion molecule 1; endothelial leukocyte adhesion molecule; endothelial leukocyte adhesion molecule 1; endothelial leukocyte adhesion molecule I; ESEL; E-selectin; LECAM2; leukocyte endothelial cell adhesion molecule 2; leukocyte-endothelial cell adhesion molecule 2; RP1-117P20.2; SELE; selectin E; selectin E (endothelial adhesion molecule 1); selectin, endothelial cell
Nom usuel E-selectin (CD62E)
Symbole de gène(s) SELE
Activité biologique Measured by its ability to support adhesion of U937 cells, a human hystiocytic lymphoma cell line.
Conjugué Unconjugated
Recombinant Recombinant
Séquence WSYNTSTEAM TYDEASAYCQ QRYTHLVAIQ NKEEIEYLNS ILSYSPSYYW IGIRKVNNVW VWVGTQKPLT EEAKNWAPGE PNNRQKDEDC VEIYIKREKD VGMWNDERCS KKKLALCYTA ACTNTSCSGH GECVETINNY TCKCDPGFSG LKCEQIVNCT ALESPEHGSL VCSHPLGNFS YNSSCSISCD RGYLPSSMET MQCMSSGEWS APIPACNVVE CDAVTNPANG FVECFQNPGS FPWNTTCTFD CEEGFELMGA QSLQCTSSGN WDNEKPTCKA VTCRAVRQPQ NGSVRCSHSP AGEFTFKSSC NFTCEEGFML QGPAQVECTT QGQWTQQIPV CEAFQCTALS NPERGYMNCL PSASGSFRYG SSCEFSCEQG FVLKGSKRLQ CGPTGEWDNE KPTCEAVRCD AVHQPPKGLV RCAHSPIGEF TYKSSCAFSC EEGFELHGST QLECTSQGQW TEEVPSCQVV KCSSLAVPGK INMSCSGEPV FGTVCKFACP EGWTLNGSAA RTCGATGHWS GLLPTCEAPT ESNIP
Contenu et stockage -20°C
Système d’expression CHO cells
Forme Lyophilized
Pureté ou qualité ≥ 95% by SDS-PAGE gel and HPLC analyses.
Afficher plus Afficher moins
Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis