missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human FAM228A (aa 107-199) Control Fragment Recombinant Protein
Click to view available options
Quantité:
100 μl
Conditionnement:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (35%), Rat (35%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-145115 (PA5-145115. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
FAM228A (Family With Sequence Similarity 228 Member A) is a Protein Coding gene. Diseases associated with FAM228A include Echolalia. An important paralog of this gene is FAM228B.
Spécification
Spécification
| Numéro d’adhésion | Q86W67 |
| Concentration | ≥5.0 mg/mL |
| À utiliser avec (application) | Blocking Assay, Control |
| Formule | 1 M urea, PBS with no preservative; pH 7.4 |
| Identification génétique (Entrez) | 653140 |
| Nom | Human FAM228A (aa 107-199) Control Fragment |
| Quantité | 100 μl |
| État réglementaire | RUO |
| Alias de gène | C2orf84; FAM228A; family with sequence similarity 228 member A; family with sequence similarity 228, member A; protein FAM228A; UPF0638 protein C2orf84 |
| Nom usuel | FAM228A |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu