Learn More
Gibco™ Human FGF-19 Recombinant Protein, PeproTech®
Code produit. 17860673
Change view
Click to view available options
Quantité:
100 μg
2 x 500 μg
25 μg
250 μg
5 μg
50 μg
500 μg
Conditionnement:
100µg
1mg
2 pièces
250µg
25µg
500µg
50µg
5µg
Code produit. Quantité unitSize
17860673 100 μg 100µg
17831773 2 x 500 μg 1mg
30087757 2 x 500 μg 2 pièces
17861483 250 μg 250µg
17871753 25 μg 25µg
17840883 500 μg 500µg
17890383 50 μg 50µg
17850703 5 μg 5µg
8 options
This item is not returnable. View return policy
Product Code. 17860673
Brand: Gibco™ 10032100UG
This item is not returnable. View return policy
Recombinant Protein
100-32-1MG will be provided as 2 x 500 μg (100-32-500UG). Recombinant Human FGF-19 is a 21.8 kDa protein containing 195 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
FGF19, Fibroblast growth factor 19, is a protein that in humans is encoded by the FGF19 gene. The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. The FGF19 gene is mapped to 11q13.3. The deduced 216-amino acid FGF19 protein contains asignal sequence and 2 cysteine residues that are conserved in the FGF family. Expression of this gene was detected only in fetal but not adult brain tissue. Synergistic interaction of the chick homolog and Wnt-8c has been shown to be required for initiation of inner ear development. FGF19 stimulates hepatic protein and glycogen synthesis but does not induce lipogenesis. The effects of FGF19 are independent of the activity of either insulin or the protein kinase Akt and, instead, are mediated through a mitogen-activated protein kinase signaling pathway that activates components of the protein translation machinery and stimulates glycogen synthase activity. The orthologous protein in mouse is FGF15, which shares about 50% amino acid identity and has similar functions. Together they are often referred to as FGF15/19.
TRUSTED_SUSTAINABILITY
Specifications
Numéro d’adhésion O95750
À utiliser avec (application) Functional Assay
Formule protein with no preservative
Identification génétique (Entrez) 9965
Poids moléculaire 21.8 kDa
Nom Human FGF-19
Quantité 100 μg
Source E. coli
État réglementaire RUO
Concentration en endotoxines <1 EU/ μg
Alias de gène FGF; Fgf15; FGF19; FGF-19; Fibroblast growth factor; fibroblast growth factor 15; fibroblast growth factor 19; UNQ334/PRO533
Nom usuel FGF19
Symbole de gène(s) FGF19
Activité biologique Determined by a cell proliferation assay using balb/c 3T3 cells. The expected ED50 for this effect is 100-150 ng/ml
Conjugué Unconjugated
Recombinant Recombinant
Séquence MRPLAFSDAG PHVHYGWGDP IRLRHLYTSG PHGLSSCFLR IRADGVVDCA RGQSAHSLLE IKAVALRTVA IKGVHSVRYL CMGADGKMQG LLQYSEEDCA FEEEIRPDGY NVYRSEKHRL PVSLSSAKQR QLYKNRGFLP LSHFLPMLPM VPEEPEDLRG HLESDMFSSP LETDSMDPFG LVTGLEAVRS PSFEK
Contenu et stockage -20°C
Système d’expression E. coli
Forme Lyophilized
Pureté ou qualité ≥ 95% by SDS-PAGE gel and HPLC analyses.