missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Human Frizzled-4 / FZD4 Protein, Fc Tag
314.52€ - 1959.64€
Spécification
Numéro d’adhésion | AAI14528 |
---|---|
Poids moléculaire | 42.9 kDa |
Source | Human Frizzled-4, Fc Tag (FZ4-H5250) is expressed from human 293 cells (HEK293). It contains AA Phe 37 - Glu 180 (Accession # AAI14528). |
Immunogène | Phe 37 - Glu 180 |
Conditions de stockage | -20°C |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15856797
|
ACROBiosystems
FZ4-H5250-100UG |
100 μg |
314.52€
100µg |
Expédition estimée: 30-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
15866797
|
ACROBiosystems
FZ4-H5250-1MG |
1 mg |
1959.64€
1mg |
Expédition estimée: 30-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
Frizzled-4 (FZD4) is also known as FzE4, CD344, which belongs to the G-protein coupled receptor Fz/Smo family. Most of frizzled receptors are coupled to the beta-catenin canonical signaling pathway, which leads to the activation of disheveled proteins, inhibition of GSK-3 kinase, nuclear accumulation of beta-catenin and activation of Wnt target genes. FZD4 contains one FZ (frizzled) domain. FZD4 may be involved in transduction and intercellular transmission of polarity information during tissue morphogenesis and/or in differentiated tissues. FZD4 interacts with MAGI3 and norrin (NDP).
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 42.9 kDa. The protein migrates as 50-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spécification
AAI14528 | |
Human Frizzled-4, Fc Tag (FZ4-H5250) is expressed from human 293 cells (HEK293). It contains AA Phe 37 - Glu 180 (Accession # AAI14528). | |
-20°C | |
Frizzled-4 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |
42.9 kDa | |
Phe 37 - Glu 180 | |
FZD4,Frizzled-4,CD344,Fz-4,hFz4,FzE4 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |