missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human FXYD2 (aa 47-66) Control Fragment Recombinant Protein

Code produit. 30204128
Click to view available options
Quantité:
100 μl
Conditionnement:
100µL
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Code produit. 30204128

Marque: Invitrogen™ RP104136

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (75%), Rat (75%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-64387 (PA5-64387. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The human FXYD2 (pronounced fix-id) gene maps to chromosome 11q23 and encodes the gamma subunit of Na/K-ATPase (NKA). FXYD2 gamma subunit modulates NKA activity by inducing ion channel activity. The mammalian FXYD family maintains Na+ and K+ gradients between the intracellular and extracellular milieus of cells in processes such as renal Na+-reabsorption, muscle contraction and neuronal excitability. FXYDs are single-span membrane proteins that share a 35 amino acid signature domain, beginning with the sequence PFXYD and containing 7 invariant and 6 conserved amino acids. Other members of the FXYD family include FXYD1 (PLM, phospholemman), FXYD3 (Mat8, mammary tumor protein 8), FXYD4 (CHIF) and FXYD5 (RIC).
TRUSTED_SUSTAINABILITY

Specifications

Numéro d’adhésion P54710
Concentration ≥5.0 mg/mL
À utiliser avec (application) Blocking Assay, Control
Formule 1 M urea, PBS with no preservative; pH 7.4
Identification génétique (Entrez) 486
Nom Human FXYD2 (aa 47-66) Control Fragment
Quantité 100 μl
État réglementaire RUO
Alias de gène ATP1C; Atp1g1; ATPase, Na+/K+ transporting, gamma 1 polypeptide; FXTD domain-containing ion transport regulator 2; FXYD domain containing ion transport regulator 2; FXYD domain-containing ion transport regulator 2; FXYD2; gamma subunit of sodium potassium ATPase; GNAKATP; HOMG2; Na(+)/K(+) ATPase subunit gamma; regulator 2; Sodium Potassium ATPase; sodium pump gamma chain; sodium/potassium-transporting ATPase gamma chain; sodium/potassium-transporting ATPase subunit gamma
Nom usuel FXYD2
Symbole de gène(s) Fxyd2
Conjugué Unconjugated
Espèces Human
Recombinant Recombinant
Marqueur de protéine His-ABP-tag
Séquence SRRFRCGGNKKRRQINEDEP
Contenu et stockage -20°C, Avoid Freeze/Thaw Cycles
Système d’expression E. coli
Forme Liquid
Pureté ou qualité >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.