missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Golgin-97 (aa 659-765) Control Fragment Recombinant Protein
Click to view available options
Quantité:
100 μl
Conditionnement:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (95%), Rat (95%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-84015 (PA5-84015. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
The Golgi apparatus, which participates in glycosylation and transport of proteins and lipids in the secretory pathway, consists of a series of stacked cisternae (flattened membrane sacs). Interactions between the Golgi and microtubules are thought to be important for the reorganization of the Golgi after it fragments during mitosis. The Golgins are a family of proteins, of which the protein encoded by this gene is a member, that are localized to the Golgi. This encoded protein is associated with Sjogren's syndrome.
Spécification
Spécification
| Numéro d’adhésion | Q92805 |
| Concentration | ≥5.0 mg/mL |
| À utiliser avec (application) | Blocking Assay, Control |
| Formule | 1 M urea, PBS with no preservative; pH 7.4 |
| Identification génétique (Entrez) | 2800 |
| Nom | Human Golgin-97 (aa 659-765) Control Fragment |
| Quantité | 100 μl |
| État réglementaire | RUO |
| Alias de gène | 0710001G09Rik; 2210418B03Rik; AW107649; gap junction protein, alpha 4, 37 kD; Golga1; golgi autoantigen, golgin subfamily a, 1; Golgi97; golgin A1; golgin subfamily A member 1; golgin-97 |
| Nom usuel | Golgin-97 |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu