missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human Gremlin-1 Recombinant Protein, PeproTech®
Click to view available options
Quantité:
10 μg
100 μg
2 x 500 μg
250 μg
50 μg
500 μg
Conditionnement:
100µg
10µg
1mg
2 pièces
250µg
500µg
50µg
Description
120-42-1MG will be provided as 2 x 500 μg (120-42-500UG). Recombinant Human Gremlin-1 is a 18.3 kDa protein containing 160 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
In Xenopus, Gremlin belongs to a novel gene family that act as BMP antagonists in embryonic explants. The human homolog of Drm/Gremlin (CKTFS1B1) is localized on human chromosome 15q13--> q15. DRM-specific mRNA is expressed in different human tissues, including brain, ovary, intestine and colon. It has been suggested that down-regulation of DRM is associated with tumor progression, and support the hypothesis that human DRM may play an important role during both neuroembryological development and carcinogenesis.
Spécification
Spécification
| Numéro d’adhésion | O60565 |
| À utiliser avec (application) | Control, Bioactivity |
| Formule | Protein with no preservative |
| Identification génétique (Entrez) | 26585 |
| Poids moléculaire | 18.3 kDa |
| Nom | Human Gremlin-1 |
| Méthode de purification | Purified |
| Quantité | 500 μg |
| Source | CHO cells |
| Conditions de stockage | -20°C |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu