missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human HOXD13 (aa 191-261) Control Fragment Recombinant Protein
Click to view available options
Quantité:
100 μl
Conditionnement:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (99%), Rat (99%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-66661 (PA5-66661. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
HOXD13 (homeobox protein D13) belongs to a class of transcription factors called homeobox proteins. In vertebrates, homeobox are found in clusters called A, B, C, and D on four separate chromosomes. Expression of homeobox proteins is spatially and temporally regulated during embryonic development. The gene encoding HOXD13 is part of the D cluster on chromosome 2. Deletions that remove the entire HOXD gene cluster or the 5' end of this cluster have been associated with severe limb and genital abnormalities. Mutations in the HOXD13 gene cause synpolydactyly.
Spécification
Spécification
| Numéro d’adhésion | P35453 |
| Concentration | ≥5.0 mg/mL |
| À utiliser avec (application) | Blocking Assay, Control |
| Formule | 1 M urea, PBS with no preservative; pH 7.4 |
| Identification génétique (Entrez) | 3239 |
| Nom | Human HOXD13 (aa 191-261) Control Fragment |
| Quantité | 100 μl |
| État réglementaire | RUO |
| Alias de gène | BDE; BDSD; homeo box 4 I; homeo box D13; homeobox D13; homeobox protein Hox-4.8; Homeobox protein Hox-4 I; homeobox protein Hox-D13; Hox-4.8; HO x 4 I; Hoxd13; SPD; SPD1; spdh |
| Nom usuel | HOXD13 |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu