missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human IFN-beta 1a Recombinant Protein

Code produit. 10669963
Click to view available options
Quantité:
1 x 10^5 unités
Conditionnement:
1 pièce
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Code produit. 10669963

Marque: Gibco™ PHC4244

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Recombinant Protein

Source: cDNA obtained from human fibroblast interferon beta 1a mRNA expressed in CHO cells. Storage: After receipt, the product may be stored at -20°C for short-term use (<6 months). For long-term storage, store the product at -70°C or below for retention of full activity. Thaw product vial by incubation in cold tap water until just thawed. The contents of the tube should be apportioned in separate tubes so that freezing and thawing is kept to a minimum. Refreezing should be done on dry ice or in a dry ice/alcohol bath. Further dilution of the product should be in buffers containing protein such as 0.1% bovine serum albumin (BSA).

Type I Interferons (IFN-alpha/beta) are produced primarily in response to viral infection by Natural IFN-producing cells (IPCs) as part of the host immune response. IFNs can also inhibit the development of tumors. IFN-beta binding results in the activation of the tyrosine kinases Jak1 and Tyk2, phosphorylation of members of the STAT family of transcription factors, and the transcription and expression of the immune response genes. More recently, several members of the toll-like receptor (TLR) family were found to stimulate the production IFN-beta. IFN-beta is currently used clinically for treatment of tumors, infections and multiple sclerosis.
TRUSTED_SUSTAINABILITY

Spécification

Numéro d’adhésion P01574
À utiliser avec (application) Bioactivity
Formule 50mM sodium acetate with 0.1% BSA and no preservative
Identification génétique (Entrez) 3456
Poids moléculaire 23 kDa
Nom Human IFN-beta 1a
Méthode de purification Purified
Quantité 1 x 10^5 unités
Conditions de stockage -80°C
État réglementaire RUO
Concentration en endotoxines <1 EU/μg
Alias de gène beta 1; fibroblast interferon; IFB; IFF; IFNB; IFNB1; IFN-beta; Interferon; Interferon beta; interferon beta 1; Interferon beta-1a; interferon, beta 1, fibroblast; interferon-beta
Nom usuel IFN beta-1a
Symbole de gène(s) IFNB1
Activité biologique EC50 ̃ 1 U/mL; determined by a cytopathic inhibition assay on human A549 lung carcinoma cells with encephalomyocarditis virus (EMCV).
Type de produit Protein
Conjugué Unconjugated
Espèces Human
Recombinant Recombinant
Contenu et stockage -80°C
Activité EC50 ~ 1 U/mL; determined by a cytopathic inhibition assay on human A549 lung carcinoma cells with encephalomyocarditis virus (EMCV).
Conditions d’expédition Dry Ice
Système d’expression CHO cells
Famille de protéines Cytokines & Receptors
Forme protéique Recombinant, Ligand
Forme Liquid
Sous-type de protéine Interferons
Catégorie de recherche Immunology, Virology, Angiogenesis
Gamme de produits Gibco
Pureté ou qualité >95%
Afficher plus Afficher moins
Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis