Learn More
Gibco™ Human IFN-beta Recombinant Protein, PeproTech®
Description
300-02BC-1MG will be provided as 2 x 500 μg (300-02BC-500UG). PeproTech's Recombinant Human IFN-beta is a 20.0 kDa protein containing 166 amino acid residues. Due to glycosylation, IFN-beta has an approximate MW of 22.3 kDa based on SDS-PAGE gel and Mass Spectrometry. *Human IFN-beta (Catalog Number 300-02BC) has replaced Human IFN-beta (Catalog Number 300-02B) This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Spécification
Spécification
| Numéro d’adhésion | P01574 |
| À utiliser avec (application) | Control, Bioactivity |
| Formule | Protein with no preservative |
| Identification génétique (Entrez) | 3456 |
| Poids moléculaire | 20 kDa |
| Nom | Human IFN-beta |
| Méthode de purification | Purified |
| Quantité | 5 μg |
| Source | CHO cells |
| Conditions de stockage | -20°C |
| Afficher plus |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.