missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human IGF-BP3 Recombinant Protein, PeproTech®

Code produit. 17820933
Click to view available options
Quantité:
1 mg
100 μg
25 μg
250 μg
5 μg
500 μg
Conditionnement:
100µg
1mg
250µg
25µg
500µg
5µg
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Code produit. 17820933

Marque: Gibco™ 10008500UG

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Recombinant Protein

Recombinant Human IGF-BP3 is a 28.8 kDa protein consisting of 264 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

IGFBP3 is a gene that belongs to the insulin-like growth factor binding protein (IGFBP) family. The gene encodes a protein that contains both an IGFBP domain and a thyroglobulin type-I domain. This protein is known to form a ternary complex with insulin-like growth factor acid-labile subunit (IGFALS) and either insulin-like growth factor (IGF) I or II. The complex circulates in the plasma and prolongs the half-life of IGFs while also altering their interaction with cell surface receptors. Various isoforms of the gene have been identified through alternate transcriptional splice variants. Diseases associated with IGFBP3 include Insulin-Like Growth Factor I and Acid-Labile Subunit Deficiency.
TRUSTED_SUSTAINABILITY

Spécification

Numéro d’adhésion P17936
À utiliser avec (application) ELISA, Functional Assay, Western Blot
Formule protein with no preservative
Identification génétique (Entrez) 3486
Poids moléculaire 28.8 kDa
Nom Human IGF-BP3
Quantité 500 μg
Source E. coli
État réglementaire RUO
Concentration en endotoxines <1 EU/ μg
Alias de gène acid stable subunit of the 140 K IGF complex; AI649005; binding protein 29; binding protein 53; BP53; BP-53; Growth hormone dependent binding protein; growth hormone-dependent binding protein; I79_011450; IBP3; IBP-3; IGF binding protein 3; IGF-binding protein 3; IGFBP; Igfbp3; IGF-BP3; IGFBP-3; IGgfbp3; Insulin like growth factor binding protein; insulin like growth factor binding protein 3; insulin-like growth factor binding protein 3; Insulin-like growth factor binding protein 3 precursor; insulin-like growth factor-binding protein (IGF-BP3); insulin-like growth factor-binding protein 3; tcag7.703
Nom usuel IGFBP3
Symbole de gène(s) IGFBP3
Activité biologique The ED50 was determined by its ability to inhibit IGF-II induced proliferation of MCF-7. The expected ED50 for this effect is ≤ 0.2 ug/ml in presence of 15 ng/ml of human IGF-II.
Conjugué Unconjugated
Recombinant Recombinant
Séquence GASSGGLGPV VRCEPCDARA LAQCAPPPAV CAELVREPGC GCCLTCALSE GQPCGIYTER CGSGLRCQPS PDEARPLQAL LDGRGLCVNA SAVSRLRAYL LPAPPAPGNA SESEEDRSAG EVESPSVSST HRVSDPKFHP LHSKIIIIKK GHAKDSQRYK VDYESQSTDT QNFSSESKRE TEYGPCRREM EDTLNHLKFL NVLSPRGVHI PNCDKKGFYK KKQCRPSKGR KRGFCWCVDK YGQPLPGYTT KGKEDVHCYS MQSK
Contenu et stockage -20°C
Système d’expression E. coli
Forme Lyophilized
Pureté ou qualité ≥ 98% by SDS-PAGE gel and HPLC analyses.
Afficher plus Afficher moins
Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis