missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human IGF-BP7, Animal-Free Recombinant Protein, PeproTech®
Click to view available options
Quantité:
1 mg
100 μg
25 μg
500 μg
Conditionnement:
100µg
1mg
25µg
500µg
Description
Recombinant Human IGF-BP7 is a 26.4 kDa protein consisting of 256 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
This gene encodes a member of the insulin-like growth factor (IGF)-binding protein (IGFBP) family. IGFBPs bind IGFs with high affinity, and regulate IGF availability in body fluids and tissues and modulate IGF binding to its receptors. This protein binds IGF-I and IGF-II with relatively low affinity, and belongs to a subfamily of low-affinity IGFBPs. It also stimulates prostacyclin production and cell adhesion. Alternatively spliced transcript variants encoding different isoforms have been described for this gene, and one variant has been associated with retinal arterial macroaneurysm.
Spécification
Spécification
| Numéro d’adhésion | Q16270 |
| À utiliser avec (application) | Functional Assay |
| Formule | protein with no preservative |
| Identification génétique (Entrez) | 3490 |
| Poids moléculaire | 26.4 kDa |
| Nom | Human IGF-BP7, Animal-Free |
| Quantité | 500 μg |
| Source | E. coli |
| État réglementaire | RUO |
| Concentration en endotoxines | <0.1 EU/ μg |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu