missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human IL-2 QuantiGlo ELISA Kit
missing translation for 'orderingAttributeHoverText'
Suffisant pour:
1 plaque à 96 puits
missing translation for 'unitSize'
1 kit
Description
The Human IL-2 QuantiGlo ELISA Kit quantifies human IL-2 in cell culture supernates (100 ul), serum (100 ul), edta plasma (100 ul), heparin plasma (100 ul).
Specifications
Specifications
| Plage de dosage | 1.70 - 1250pg/mL |
| Sensibilité du dosage | 0.25pg/mL |
| Conjugué | HRP |
| Type d’échantillon | Cell Culture Supernatant, Plasma (EDTA), Plasma (Heparin), Serum |
| Spécificité | Natural and Recombinant Human IL-2 |
| Suffisant pour | 1 plaque à 96 puits |
| À utiliser avec (application) | ELISA |
| Identification génétique (Entrez) | 3558 |
| Volume d’échantillon | Cell Culture Supernates (100uL),Serum (100uL),EDTA Plasma (100uL),Heparin Plasma (100uL) |
| Conditions de stockage | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction