missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human IL-33 Recombinant Protein, PeproTech®

Code produit. 17850603
Click to view available options
Quantité:
1 mg
10 μg
100 μg
2 μg
250 μg
50 μg
500 μg
Conditionnement:
100µg
10µg
1mg
250µg
2µg
500µg
50µg
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Code produit. 17850603

Marque: Gibco™ 20033100UG

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Recombinant Protein

Recombinant Human IL-33 is a 17.9 kDa protein containing 159 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

IL-33 (Interleukin-33) is a 270 amino acid, highly divergent protein belonging to the IL-1 family with an IL-1-like C-terminal domain. IL-33 is a dual function protein that may function both as a proinflammatory cytokine and an intracellular nuclear factor with transcriptional regulatory properties. IL-33 binds to and signals through IL1RL1/ST2 and its stimulation recruits MYD88, IRAK1, IRAK4, and TRAF6. IL-33 activates NF-kappaB and MAP kinases, and drives production of TH2-associated cytokines from in vitro polarized TH2 cells. In vivo, IL-33 induces the expression of IL-4, IL-5, and IL-13 and leads to severe pathological changes in mucosal organs. IL-33 is proteolytically converted to a mature form by CASP1 and is highly expressed in high endothelial venules found in tonsils, Peyer's patches and mesenteric lymph nodes and is almost undetectable in placenta. Prolonged IL-33 treatment of mice led to the development of eosinophilia, splenomegaly, and severe pathological changes in mucosal organs such as lungs, esophagus and small intestine. Recent experiments have shown that IL-33 can also co-localize with heterochromatin and possesses transcriptional repressor activities, indicating that IL-33 may function as both a proinflammatory cytokine and an intracellular nuclear factor with transcriptional regulatory properties. Despite its predicted molecular weight, IL-33 will often run at higher molecular weight in SDS-PAGE. Studies have shown that IL-33 can also co-localize with heterochromatin and possesses transcriptional repressor activities, indicating that IL-33 may function as both a proinflammatory cytokine, and an intracellular nuclear factor with transcriptional regulatory properties.
TRUSTED_SUSTAINABILITY

Spécification

Numéro d’adhésion O95760, Q01638
À utiliser avec (application) ELISA, Functional Assay, Western Blot
Formule protein with no preservative
Identification génétique (Entrez) 90865, 9173
Poids moléculaire 17.9 kDa
Nom Human IL-33
Quantité 100 μg
Source E. coli
État réglementaire RUO
Concentration en endotoxines <1 EU/ μg
Alias de gène 9230117N10Rik; C9orf26; DVS27; DVS27-related protein; H-IL-33; IL1F11; IL-1F11; Il33; IL-33; ILN; Interleukin; interleukin 33; interleukin-1 family member 11; Interleukin33; Interleukin-33; Interleukin-33 (109-270); Interleukin-33 (95-270); Interleukin-33 (99-270); Interleukin-33(102-266); Interleukin-33(109-266); NFEHEV; NFHEV; NF-HEV; nuclear factor for high endothelial venules; nuclear factor from high endothelial venules; RGD1311155; RP11-575C20.2
Nom usuel IL-33
Symbole de gène(s) IL33
Activité biologique The ED50 was determined by the dose-dependent stimulation of the proliferation of murine D10S cells is ≤ 0.05 ng/ml, corresponding to a specific activity of ≥ 2 x 10^7 units/mg
Conjugué Unconjugated
Recombinant Recombinant
Séquence SITGISPITE YLASLSTYND QSITFALEDE SYEIYVEDLK KDEKKDKVLL SYYESQHPSN ESGDGVDGKM LMVTLSPTKD FWLHANNKEH SVELHKCEKP LPDQAFFVLH NMHSNCVSFE CKTDPGVFIG VKDNHLALIK VDSSENLCTE NILFKLSET
Contenu et stockage -20°C
Système d’expression E. coli
Forme Lyophilized
Pureté ou qualité ≥ 98% by SDS-PAGE gel and HPLC analyses.
Afficher plus Afficher moins
Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis