missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human IL-4 Receptor alpha (soluble) Recombinant Protein, PeproTech®

Code produit. p-7246271
Click to view available options
Quantité:
1 mg
100 μg
20 μg
250 μg
5 μg
500 μg
Conditionnement:
100µg
1mg
20µg
250µg
500µg
5µg
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Code produit. 17899902

Marque: Gibco™ 20004RC100UG

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Recombinant Protein

PeproTech's CHO cell-derived Recombinant Human sIL-4 Receptor alpha is a 23.9 kDa glycoprotein corresponding to 209 amino acid residues of the extracellular domain of IL-4Ralpha. As a result of glycosylation, Recombinant Human sIL-4 Receptor alpha migrates with an apparent molecular mass of approximately 50-65 kDa by SDS-PAGE gel, under reducing conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

IL4 Receptor alpha (IL4RA) is the alpha chain of the interleukin 4 receptor which binds to both interleukin 4 and interleukin 13 to regulate IgE production, chemokine and mucus production at sites of allergic inflammmation. The IL4 response is involved in promoting Th2 differentiation. The secreted extracellular domain of IL4R alpha, called sIL4R alpha, can inhibit IL4-mediated cell proliferation and IL5 upregulation by T-cells. It has no signaling abilities. Allelic variations in the IL4RA gene have been associated with atopy, a condition that can manifest itself as allergic rhinitis, sinusitus, asthma, or eczema.
TRUSTED_SUSTAINABILITY

Spécification

Numéro d’adhésion P24394
À utiliser avec (application) ELISA, Functional Assay, Western Blot
Formule protein with no preservative
Identification génétique (Entrez) 3566
Poids moléculaire 23.9 kDa
Nom Human IL-4 Receptor alpha (soluble)
Quantité 100 μg
Source CHO cells
État réglementaire RUO
Concentration en endotoxines <1 EU/ μg
Alias de gène 582J2.1; CD124; il-4 receptor; IL-4 receptor alpha chain; IL-4 receptor subunit alpha; IL-4-binding protein; IL4-BP; Il4r; IL4R nirs variant 1; IL-4 R subunit alpha; IL4RA; IL-4 RA; IL-4 R-alpha; interleukin 4 receptor; interleukin 4 receptor, alpha; interleukin-4 receptor alpha chain; interleukin-4 receptor subunit alpha; Soluble IL-4 receptor subunit alpha; Soluble interleukin-4 receptor subunit alpha
Nom usuel IL4R
Symbole de gène(s) IL4R
Activité biologique The ED50 was determined by its ability to inhibit the IL-4 dependent proliferation of human TF-1 cells is ≤ 5.0 ng/ml (in the presence of 0.5 ng/ml of IL-4), corresponding to a specific activity of ≥ 2 x 10^5 units/mg
Conjugué Unconjugated
Recombinant Recombinant
Séquence GNMKVLQEPT CVSDYMSIST CEWKMNGPTN CSTELRLLYQ LVFLLSEAHT CIPENNGGAG CVCHLLMDDV VSADNYTLDL WAGQQLLWKG SFKPSEHVKP RAPGNLTVHT NVSDTLLLTW SNPYPPDNYL YNHLTYAVNI WSENDPADFR IYNVTYLEPS LRIAASTLKS GISYRARVRA WAQCYNTTWS EWSPSTKWHN SYREPFEQH
Contenu et stockage -20°C
Système d’expression CHO cells
Forme Lyophilized
Pureté ou qualité ≥ 95% by SDS-PAGE gel and HPLC analyses.
Afficher plus Afficher moins
Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis