missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human MAP2 (aa 1540-1674) Control Fragment Recombinant Protein
Click to view available options
Quantité:
100 μl
Conditionnement:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (96%), Rat (96%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-110744 (PA5-110744. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
MAP2 (Microtubule Associated Protein 2) exists in two high molecular weight forms (MAP2a & MAP2b) and a low molecular weight form. The expression of MAP2 is developmentally regulated and its multiple forms arise by alternative splicing of a single gene. MAP2 is involved in microtubule assembly, which is an essential step in neurogenesis. The products of similar genes in rat and mouse are neuron-specific cytoskeletal proteins that are enriched in dendrites, implicating a role in determining and stabilizing dendritic shape during neuron development. MAP2 is useful in studies of neuron structure in normal and malignant brain tissue as well as in degenerative diseases such as Alzheimer's Disease (AD).
Spécification
Spécification
| Numéro d’adhésion | P11137 |
| Concentration | ≥5.0 mg/mL |
| À utiliser avec (application) | Blocking Assay, Control |
| Formule | 1 M urea, PBS with no preservative; pH 7.4 |
| Identification génétique (Entrez) | 4133 |
| Nom | Human MAP2 (aa 1540-1674) Control Fragment |
| Quantité | 100 μl |
| État réglementaire | RUO |
| Alias de gène | a730034c02; AC091465.1; DKFZp686I2148; G1-397-34; MAP 2; Map2; MAP-2; map2.S; map2a; map-2 A; map2ab; MAP2B; MAP2C; MAP2R; microtubule associated protein 2; microtubule associated protein 2 S homeolog; microtubule associated protein 2; microtubule-associated protein 2; microtubule associated protein 2 C; microtubule-associated protein 2; microtubule-associated protein-2; Mtap2; Mtap-2; OTTHUMP00000163916; OTTHUMP00000208143; repro4; XELAEV_18047318mg; xmap2 |
| Nom usuel | MAP2 |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu