missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human MIA Recombinant Protein, PeproTech®

Code produit. 17891023
Click to view available options
Quantité:
1 mg
100 μg
20 μg
250 μg
5 μg
500 μg
Conditionnement:
100µg
1mg
20µg
250µg
500µg
5µg
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Code produit. 17891023

Marque: Gibco™ 1300120UG

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Recombinant Protein

Recombinant Human MIA is a 12.2 kDa globular protein containing 108 amino acid residues, including two intramolecular disulfide bonds. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Tumorigenesis is a process that is mediated by a network of membrane, cytosolic and nuclear associated factors, which regulate proliferation and cell-matrix interaction through signaling cascades. The phenotype of malignant melanomas in vivo depends on the global expression of stimulatory or inhibitory factors generated in both the tumors cells and their environment. One example, Melanoma inhibitory activity (cartilage-derived retinoic acid-sensitive protein (CD-RAP), MIA) is a Src homology 3 (SH3)-like domain containing protein that is secreted from chondrocytes and malignant melanoma cells. MIA is translated as a 131-amino acid pro-form and processed into a mature 107-amino acid protein after cleavage of a secretion signal. MIA is expressed during chondrogenesis and in mature chondrocytes, suggesting that MIA is necessary for normal cartilage cell phenotype. MIA mRNA is present in carcinomas of the colon, ovary, kidney, and head/neck, and may represent a marker to monitor melanomic activity.
TRUSTED_SUSTAINABILITY

Spécification

Numéro d’adhésion Q16674
À utiliser avec (application) ELISA, Functional Assay, Western Blot
Formule protein with no preservative
Identification génétique (Entrez) 8190
Poids moléculaire 12.2 kDa
Nom Human MIA
Quantité 20 μg
Source E. coli
État réglementaire RUO
Concentration en endotoxines <1 EU/ μg
Alias de gène cartilage derived retinoic acid sensitive protein; cartilage-derived retinoic acid-sensitive protein; Cdrap; CD-RAP; melanoma inhibitory activity; melanoma inhibitory activity 1; melanoma inhibitory activity protein; melanoma inhibitory activity/condrocyte-derived retinoic acid sensitive protein homolog (MIA/CD-RAP); melanoma-derived growth regulatory protein; melanoma-inhibitory-activity; MIA; Mia1
Nom usuel MIA
Symbole de gène(s) MIA
Activité biologique The ED50 was determined by the dose-dependant proliferation of the human A375 melanoma cell line. The expected ED50 for this effect is 4-6 ug/ml
Conjugué Unconjugated
Recombinant Recombinant
Séquence MGPMPKLADR KLCADQECSH PISMAVALQD YMAPDCRFLT IHRGQVVYVF SKLKGRGRLF WGGSVQGDYY GDLAARLGYF PSSIVREDQT LKPGKVDVKT DKWDFYCQ
Contenu et stockage -20°C
Système d’expression E. coli
Forme Lyophilized
Pureté ou qualité ≥ 98% by SDS-PAGE gel and HPLC analyses.
Afficher plus Afficher moins
Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis