missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human MORF4L1 Partial ORF (NP_006782.1, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marque: Abnova™ H00010933-Q01.10ug
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Sequence: MAPKQDPKPKFQEGERVLCFHGPLLYEAKCVKVAIKDKQVKYFIHYSGWNKNWDEWVPESRVLKYVDTNLQKQRELQKANQEQYAEGKMRGAAPGKKTSGSpécification
NP_006782.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAPKQDPKPKFQEGERVLCFHGPLLYEAKCVKVAIKDKQVKYFIHYSGWNKNWDEWVPESRVLKYVDTNLQKQRELQKANQEQYAEGKMRGAAPGKKTSG | |
RUO | |
MORF4L1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
10933 | |
MORF4L1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
Eaf3/FWP006/HsT17725/MGC10631/MORFRG15/MRG15/S863-6 | |
MORF4L1 | |
Recombinant | |
wheat germ expression system |