Learn More
Gibco™ Human/Mouse/Rat BMP-2 Recombinant Protein, PeproTech®
Description
Recombinant Human/Murine/Rat BMP-2 derived from CHO cells is a homodimeric glycoprotein that consists of two 114 amino acid polypeptide chains linked by a single disulfide bond. Due to glycosylation, CHO cell-derived Human/Murine/Rat BMP-2 migrates at an apparent molecular weight of approximately 28-29 kDa by SDS-PAGE analysis under non-reducing conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Spécification
Spécification
| Numéro d’adhésion | P12643, P21274, P49001 |
| À utiliser avec (application) | Functional Assay |
| Formule | protein with no preservative |
| Identification génétique (Entrez) | 12156, 29373, 650 |
| Poids moléculaire | 25.8 kDa |
| Nom | Human/Mouse/Rat BMP-2 |
| Quantité | 500 μg |
| Source | CHO cells |
| État réglementaire | RUO |
| Concentration en endotoxines | <1 EU/ μg |
| Afficher plus |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.