missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human NENF (aa 45-104) Control Fragment Recombinant Protein
Click to view available options
Quantité:
100 μl
Conditionnement:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (97%), Rat (97%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-55410 (PA5-55410. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
NENF is a secreted protein that is expressed in neurons but not glial cells of the brain. This protein has neurotrophic activity in primary cultured mouse neurons but not mitogenic activity in primary cultured mouse astrocytes. NENF activated mitogen-activated protein (MAP) and phosphotidylinositol-3 kinase pathways and could be inhibited by pertussis toxin but not receptor tyrosine kinases, suggesting that NENF acts through a Gi/Go-protein-coupled receptor.
Spécification
Spécification
| Numéro d’adhésion | Q9UMX5 |
| Concentration | ≥5.0 mg/mL |
| À utiliser avec (application) | Blocking Assay, Control |
| Formule | 1 M urea, PBS with no preservative; pH 7.4 |
| Identification génétique (Entrez) | 29937 |
| Nom | Human NENF (aa 45-104) Control Fragment |
| Quantité | 100 μl |
| État réglementaire | RUO |
| Alias de gène | 1110060M21Rik; cell growth-inhibiting protein 47; cell immortalization-related protein 2; CIR2; NENF; nenf {ECO:0000250; Neudesin; neudesin neurotrophic factor; neuron derived neurotrophic factor; neuron-derived neurotrophic factor; Protein GIG47; SCIRP10; SCIRP10-related protein; secreted protein of unknown function; sp2; SP2 protein; Spinal cord injury related protein 10; spinal cord injury-related protein 10; SPUF; SPUF protein; UniProtKB:Q9CQ45}; wu:fa11a04; wu:fb73f07; zgc:112953; zgc:112953 protein; zgc:152703 |
| Nom usuel | NENF |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu