missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Human OX40 / TNFRSF4 / CD134 Protein, Fc Tag
Marque: ACROBiosystems OX0-H5255-1MG
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Tumor necrosis factor receptor superfamily member 4 (TNFRSF4) is also known as ACT35 antigen, OX40L receptor, TAX transcriptionally-activated glycoprotein 1 receptor, CD antigen CD134, OX40. OX40 / TNFRSF4 contains four TNFR-Cys repeats. TNFRSF4 is receptor for TNFSF4 / OX40L / GP34 and can interacts with TRAF2, TRAF3 and TRAF5.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 46.8 kDa. The protein migrates as 50-67 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spécification
NP_003318 | |
1 mg | |
Leu 29 - Ala 216 | |
TNFRSF4,OX40,CD134,OX40L receptor,ACT35,TXGP1L | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
46.8 kDa | |
Human O x 40, Fc Tag (O x 0-H5255) is expressed from human 293 cells (HEK293). It contains AA Leu 29 - Ala 216 (Accession # NP_003318). | |
-20°C | |
OX40 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Immune Checkpoint Proteins |