Learn More
STEMCELL Technologies Human pan IFN-alpha ELISA Kit
Code produit. 17198001
Encompass_Preferred
Click to view available options
Quantité:
10 plaques
2 plaques
Conditionnement:
10 pièces
2 pièces
Code produit. 17198001
Marque: STEMCELL Technologies 02001
For detection and measurement of pan human interferon-alpha
  • The Human pan Interferon-alpha (IFN-α) ELISA Kit is designed for the quantitative detection and measurement of human IFN-α in biological fluids such as serum, plasma, and cell culture supernatants
  • IFN-α is primarily produced by macrophages resulting from viral infections and activates anti-viral and immunomodulatory activities in target cells
  • This kit is specific for subtypes 1/13, 2, 4, 5, 6, 7, 8, 10, 14, 16, and 17
  • The assay is based on the sandwich ELISA method, in which samples are added to ELISA strip plates pre-coated with capture antibodies specific for the cytokine
  • The captured cytokine is detected by addition of a biotinylated detection antibody, followed by streptavidin-horseradish peroxidase, which binds the biotinylated antibody
  • Addition of the chromogenic enzyme substrate 3,3',5,5' tetramethylbenzidine (TMB) results in a colored product with an intensity directly proportional to the concentration of cytokine in the sample
Spécification
Description Dendritic Cells; Macrophages
À utiliser avec (application) ELISA
Quantité 10 plaques
Correction du contenu d'un produit
Le programme Encompass de Fisher Scientific propose des articles qui ne font pas partie de notre portefeuille de distribution. Ces produits n'ont généralement pas de photos ou de descriptions détaillées. Cependant, nous nous engageons à améliorer votre expérience d'achat. Veuillez utiliser le formulaire ci-dessous pour nous faire part de vos commentaires sur le contenu de ce produit.