Learn More
Abnova™ Human PART1 Full-length ORF (AAH69304.1, 1 a.a. - 59 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marque: Abnova™ H00025859-P01.10ug
Informations supplémentaires : Poids : 0.00010kg
Description
This gene is induced by androgen in prostate adenocarcinoma cells. Multiple alternatively transcript variants have been described for this gene, none of which are predicted to encode a protein product. [provided by RefSeq]
Sequence: MQCQLFRTETSKAVSELNYDYICIKAGTGRPQGTPTIGLVLLVRWAIIYETELQSQPITSpécification
AAH69304.1 | |
Liquid | |
25859 | |
PART1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MQCQLFRTETSKAVSELNYDYICIKAGTGRPQGTPTIGLVLLVRWAIIYETELQSQPIT | |
RUO | |
PART1 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
33kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp586D0823 | |
PART1 | |
Yes | |
wheat germ expression system |