missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PD-1 His-tag Fc Chimera Recombinant Protein

Code produit. 17134203
Click to view available options
Quantité:
50 μg
Conditionnement:
50µg
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Code produit. 17134203

Marque: Invitrogen™ A42530

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Recombinant Protein

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/mL or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Cell-mediated immune responses are initiated by T lymphocytes that are themselves stimulated by cognate peptides bound to MHC molecules on antig en-presenting cells (APC). T-cell activation is generally self-limited as activated T cells express receptors such as PD-1 (also known as PDCD-1) that mediate inhibitory signals from the APC. PD-1 can bind two different but related ligands, PDL-1 and PDL-2. Upon binding to either of these ligands, signals generated by PD-1 inhibit the activation of the immune response in the absence of 'danger signals' such as LPS or other molecules associated with bacteria or other pathogens. Evidence for this is seen in PD1-null mice who exhibit hyperactivated immune systems and autoimmune diseases. Despite its predicted molecular weight, PD-1 often migrates at higher molecular weight in SDS-PAGE.
TRUSTED_SUSTAINABILITY

Spécification

Numéro d’adhésion Q15116
À utiliser avec (application) Control, ELISA Standard, Binding Assay
Formule PBS with 5% mannitol, 5% trehalose, 0.01% Tween 80 and no preservative; pH 7.4
Identification génétique (Entrez) 5133
Poids moléculaire 44 kDa
Nom Human PD-1 His-tag Fc Chimera
Plage de pH 7.4
Méthode de purification Purified
Quantité 50 μg
Source HEK293 cells
Conditions de stockage -20°C or -80°C if preferred
État réglementaire RUO
Concentration en endotoxines <1.0 EU/μg
Alias de gène CD279; EGK_05005; hPD1; hPD-1; hPD-l; hSLE1; Ly101; mPD-1; PD1; PD-1; Pdc1; Pdcd1; programmed cell death 1; programmed cell death 1 protein; programmed cell death protein 1; programmed cell death protein 1-like; programmed death 1; Protein PD1; protein PD-1; sCD279; SLEB2; soluble CD279; systemic lupus erythematosus susceptibility 2
Nom usuel PD-1 (CD279)
Symbole de gène(s) Pdcd1
Activité biologique Measured by its binding ability in a functional ELISA. Immobilized recombinant human PD-L2 (at 1 μg/mL (100 μL/well) can bind human PD1/Fc chimera with a linear range of 7.8-1000 ng/mL)
Type de produit Protein
Conjugué Unconjugated
Espèces Human
Recombinant Recombinant
Marqueur de protéine Fc-tag, His-tag
Séquence Human PD1, amino acids Met1-Gln167 (Accession # NP_005009.2) with a C-terminal His-tagged human IgG1 Fc-tag
Contenu et stockage Includes 1 vial of lyophilized protein. Store at -20 °C.
Activité Measured by its binding ability in a functional ELISA. Immobilized recombinant human PD-L2 at 1 μg/mL (100 μL/well) can bind human PD1/Fc chimera with a linear range of 7.8-1000 ng/mL.
Niveau d’endotoxines < 1 EU/μg
Système d’expression HEK293 cells
Famille de protéines CD Proteins
Forme Lyophilized
Sous-type de protéine Other Proteins
Catégorie de recherche Apoptosis, Immunology, Inflammation
Pureté ou qualité >97% by SDS-PAGE
Protéine programmed cell death 1
Afficher plus Afficher moins
Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis