missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human PPP1R14C Full-length ORF (AAH15773.1, 1 a.a. - 165 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marque: Abnova™ H00081706-P01.25ug
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Sequence: MSVATGSSEAAGGASGGGARVFFQSPRGGAGGSPGSSSGSGSSREDSAPVATAAAAGQVQQQQQRRHQQGKVTVKYDRKELRKRLVLEEWIVEQLGQLYGCEEEEMPEVEIDIDDLLDADSDEERASKLQEALVDCYKPTEEFIKELLSRIRGMRKLSPPQKKSVSpécification
AAH15773.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
44.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CPI17-like/KEPI/NY-BR-81 | |
PPP1R14C | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
81706 | |
PPP1R14C (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSVATGSSEAAGGASGGGARVFFQSPRGGAGGSPGSSSGSGSSREDSAPVATAAAAGQVQQQQQRRHQQGKVTVKYDRKELRKRLVLEEWIVEQLGQLYGCEEEEMPEVEIDIDDLLDADSDEERASKLQEALVDCYKPTEEFIKELLSRIRGMRKLSPPQKKSV | |
RUO | |
PPP1R14C | |
Wheat Germ (in vitro) | |
GST | |
Liquid |