Learn More
STEMCELL Technologies Human Recombinant BDNF
Code produit. p-7217729
Encompass_Preferred
Click to view available options
Quantité:
10 μg
100 μg
1 000 μg
500 μg
Conditionnement:
1000µg
500µg
Code produit. 17159321
Marque: STEMCELL Technologies 78005
Brain-derived neurotrophic factor
  • Brain-derived neurotrophic factor (BDNF), like nerve growth factor (NGF), neurotrophin-3 (NT-3), and neurotrophin-4 (NT-4), is a member of the NGF family of neurotrophins, which are required for the differentiation and survival of specific neuronal subpopulations in both the central and the peripheral nervous systems
  • BDNF binds with high affinity to the tropomyosin receptor kinase B (TrkB), and activates AKT and ERK pathways
  • It is expressed in the hippocampus, cortex, and synapses of the basal forebrain
  • BDNF acts as a survival factor for human embryonic stem cells when plated on either feeder cells or Corning™ Matrigel™
Spécification
Nom Abrineurin, ANON2, BULN2, Neurotrophin, MGC34632
Quantité 10 μg
Type de cellule Neural Cells, PSC-Derived; Neural Stem and Progenitor Cells; Neurons
Pureté ou qualité ≥95%