Learn More
STEMCELL Technologies Human Recombinant CNTF
Code produit. p-7217756
Encompass_Preferred
Click to view available options
Quantité:
10 μg
50 μg
Conditionnement:
10µg
Code produit. 17129341
Marque: STEMCELL Technologies 78010
Ciliary neurotrophic factor
  • Ciliary neurotrophic factor (CNTF) is a neurotrophic factor that belongs to the four-helix bundle cytokine family and is structurally related to interleukin 6 (IL-6), interleukin 11 (IL-11), leukemia inhibitory factor (LIF), and oncostatin M (OSM)
  • CNTF binds to its receptor CNFTRα and induces formation of a heterodimer of the signal-transducing IL-6 receptor gp130 and LIF receptor (LIFR)-β, which triggers JAK/STAT, ERK, and the PI3K signaling cascades
  • CNTF plays an important role in neurogenesis and the differentiation of neural stem cells and has been suggested to possess a therapeutic role in treating neurological disorders
  • CNTF has also been shown to protect rod photoreceptors from light-induced damage and to have therapeutic effects on retinal degenerative diseases caused by genetic defects or damage induced by toxins, autoantibodies, or strong light
Spécification
Nom Ciliary neurotrophic factor
Quantité 10 μg
Type de cellule Neural Cells, PSC-Derived; Neural Stem and Progenitor Cells; Neurons
Pureté ou qualité ≥95%
Correction du contenu d'un produit
Le programme Encompass de Fisher Scientific propose des articles qui ne font pas partie de notre portefeuille de distribution. Ces produits n'ont généralement pas de photos ou de descriptions détaillées. Cependant, nous nous engageons à améliorer votre expérience d'achat. Veuillez utiliser le formulaire ci-dessous pour nous faire part de vos commentaires sur le contenu de ce produit.