Learn More
STEMCELL Technologies Human Recombinant FGF-10 (KGF-2), ACF
Code produit. 17139751
Encompass_Preferred
Click to view available options
Quantité:
100 μg
1 000 μg
Conditionnement:
100µg
Code produit. 17139751
Marque: STEMCELL Technologies 78173.1
Fibroblast growth factor 10, animal component-free
  • Fibroblast growth factor 10 (FGF-10) is a member of the fibroblast growth factor (FGF) family which is predominantly expressed by mesenchymal fibroblasts during embryonic development
  • It binds with high affinity to fibroblast growth factor receptor 2-IIIb (FGFR2-IIIb), and also has a weaker affinity for FGFR1-IIIb
  • FGF-10 and FGF-7 have similar receptor binding properties and target cell specificities but are differentially regulated by components of the extracellular matrix
  • FGF-10 also has a role in mobilization and proliferation of lung-resident mesenchymal stem cells (MSCs) and protection and repair against acute lung injury and endodermal differentiation of human pluripotent stem cells to insulin-producing pancreatic-like cells
  • This product is animal component-free
Spécification
Nom Fibroblast growth factor 10, Keratinocyte growth factor 2
Quantité 100 μg
Type de cellule Endoderm, PSC-Derived; Hematopoietic Stem and Progenitor Cells; Keratinocytes; Mesenchymal Stem and Progenitor Cells; Neural Cells, PSC-Derived; Neural Stem and Progenitor Cells; Pluripotent Stem Cells; Prostate Cells
Pureté ou qualité ≥95%
Correction du contenu d'un produit
Le programme Encompass de Fisher Scientific propose des articles qui ne font pas partie de notre portefeuille de distribution. Ces produits n'ont généralement pas de photos ou de descriptions détaillées. Cependant, nous nous engageons à améliorer votre expérience d'achat. Veuillez utiliser le formulaire ci-dessous pour nous faire part de vos commentaires sur le contenu de ce produit.