Learn More
STEMCELL Technologies Human Recombinant FGF-10 (KGF-2), ACF
Click to view available options
Quantité:
100 μg
1 000 μg
Conditionnement:
100µg
Description
- Fibroblast growth factor 10 (FGF-10) is a member of the fibroblast growth factor (FGF) family which is predominantly expressed by mesenchymal fibroblasts during embryonic development
- It binds with high affinity to fibroblast growth factor receptor 2-IIIb (FGFR2-IIIb), and also has a weaker affinity for FGFR1-IIIb
- FGF-10 and FGF-7 have similar receptor binding properties and target cell specificities but are differentially regulated by components of the extracellular matrix
- FGF-10 also has a role in mobilization and proliferation of lung-resident mesenchymal stem cells (MSCs) and protection and repair against acute lung injury and endodermal differentiation of human pluripotent stem cells to insulin-producing pancreatic-like cells
- This product is animal component-free
Spécification
Spécification
| Nom | Fibroblast growth factor 10, Keratinocyte growth factor 2 |
| Quantité | 100 μg |
| Type de cellule | Endoderm, PSC-Derived; Hematopoietic Stem and Progenitor Cells; Keratinocytes; Mesenchymal Stem and Progenitor Cells; Neural Cells, PSC-Derived; Neural Stem and Progenitor Cells; Pluripotent Stem Cells; Prostate Cells |
| Pureté ou qualité | ≥95% |