Learn More
STEMCELL Technologies Human Recombinant FGF-7 (KGF)
Click to view available options
Quantité:
10 μg
1 000 μg
50 μg
Conditionnement:
10µg
1mg
50µg
Description
- Fibroblast growth factor 7 (FGF-7) is a member of the FGF family, and acts exclusively through a subset of FGF receptor isoforms expressed predominantly by epithelial cells (Finch & Rubin)
- FGF-7 seems to act specifically on epithelial cells and stimulates proliferation, migration, and differentiation of these cells, and also participates in epithelial protection and repair both in vitro and in vivo (Finch & Rubin; Werner)
- In contrast, FGF-7 is produced solely by cells of mesenchymal origin, and functions as a paracrine mediator of mesenchymal-epithelial communication (Rubin et al.) FGF-7 has also been shown to supplement several wound-healing properties of bioengineered skin (Erdag et al.) and to induce autophagy in human keratinocytes (Belleudi et al.) Additionally, FGF-7 has a role in pluripotent stem cell differentiation to endodermal pancreatic-like insulin-producing cells and thymic epithelial cells (Inami et al.; Niu et al.)
Specifications
Specifications
| Nom | Fibroblast growth factor-7, HBGF-7, Heparin-binding growth factor 7, Keratinocyte growth factor |
| Quantité | 1 000 μg |
| Type de cellule | Endoderm, PSC-Derived; Hematopoietic Stem and Progenitor Cells; Keratinocytes; Mammary Cells; Mesenchymal Stem and Progenitor Cells; Neural Cells, PSC-Derived; Neural Stem and Progenitor Cells; Pluripotent Stem Cells; Prostate Cells |
| Pureté ou qualité | ≥95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction