Learn More
STEMCELL Technologies Human Recombinant Fractalkine (CX3CL1)
Click to view available options
Quantité:
1 000 μg
25 μg
Conditionnement:
1000µg
25µg
Description
- Fractalkine (CX3CL1) is a unique chemokine belonging to the CX3C family, and is characterized by a C-X3-C cysteine motif within the chemokine domain, near the amino terminus of the protein
- The chemokine domain is connected to an extended mucin-like stalk, followed by a transmembrane region, and a C-terminal intracellular domain
- The protein signals through interaction with a single receptor, CX3CR1, expressed on monocytes, natural killer cells, T cells, microglia, and smooth muscle cells
- Fractalkine is upregulated in endothelial cells by inflammatory signals and is synthesized as a membrane-bound molecule that mediates cell migration and adhesion
- Cleavage at the base of the stalk by metalloproteinases generates a soluble chemokine, which functions as a potent chemoattractant of target cells
Spécification
Spécification
| Nom | C3Xkine, Chemokine (C-X3-C motif) ligand 1, FKN, Neurotactin, NTN, NTT, SCYD1, Small inducible cytokine subfamily D member 1 |
| Quantité | 1 000 μg |
| Type de cellule | Dendritic Cells; Endothelial Cells; Macrophages; Monocytes; NK Cells; T Cells |
| Pureté ou qualité | ≥95% |
Correction du contenu d'un produit
Le programme Encompass de Fisher Scientific propose des articles qui ne font pas partie de notre portefeuille de distribution. Ces produits n'ont généralement pas de photos ou de descriptions détaillées. Cependant, nous nous engageons à améliorer votre expérience d'achat. Veuillez utiliser le formulaire ci-dessous pour nous faire part de vos commentaires sur le contenu de ce produit.