Learn More
STEMCELL Technologies Human Recombinant HBEGF
Click to view available options
Quantité:
10 μg
50 μg
Conditionnement:
10µg
Description
- Heparin-binding epidermal growth factor (EGF)-like growth factor (HBEGF) is a member of the EGF family (Nishi & Klagsbrun)
- HBEGF promotes blastocyst adhesion to the uterine wall (Iwamoto & Mekada)
- It also plays a role in smooth muscle cell hyperplasia and brain injury (Nishi & Klagsburn)
- HBEGF produced by CD4+ T cells promotes wound healing by stimulating migration and proliferation of keratinocytes, fibroblasts, and smooth muscle cells (Blotnick et al.) It binds to EGFR, ErbB4, ErbB2, and ErbB3, activating the PI3K/AKT signaling cascade (Iwamoto & Mekada)
- HBEGF is produced in a variety of cells, where it contributes to physiological and pathological processes
- HBEGF is overexpressed in ovarian, breast, gastric, colorectal, pancreatic, and endometrial cancers, which likely contributes to pathogenesis (Miyata et al.)
Spécification
Spécification
| Nom | Diphtheria toxin receptor, DTR, HBEGF, Heparin binding EGF-like growth factor |
| Quantité | 10 μg |
| Type de cellule | Fibroblasts; Keratinocytes; Smooth Muscle; T Cells, CD4+ |
| Pureté ou qualité | ≥95% |