Learn More
STEMCELL Technologies Human Recombinant Heregulin-beta 1
Click to view available options
Quantité:
10 μg
1 000 μg
50 μg
Description
- Heregulin-beta 1 also known as neuregulin-1 (NRG-1) is a member of the epidermal growth factor (EGF) family of growth factors and acts as a ligand for ErbB family receptor tyrosine kinases
- Heregulin/neuregulin is a family of structurally related polypeptide growth factors derived from alternatively spliced genes (NRG1, NRG2, NRG3, and NRG4)
- Heregulin-beta 1 plays an important role during the development of the nervous system, heart, and mammary glands
- Heregulin-beta 1 is expressed in neuronal cells, and modulates cell growth and differentiation of the cells during development and wound healing
- It has been implicated through in vivo and in vitro studies that heregulin-beta 1/ErbB signaling is crucial for multiple aspects of cardiovascular development and protects the heart from ischemic injury
- Heregulin-beta 1 also promotes invasiveness and metastasis of breast cancer cells
Spécification
Spécification
| Nom | Acetylcholine receptor-inducing activity, ARIA, Breast cancer cell differentiation factor p45, Glial growth factor, Heregulin Neu differentiation factor, HRG, HRG1, HRG1-beta 1, Neuregulin-1, NRG1, NRG1-beta 1, Sensory and motor neuron-derived factor |
| Quantité | 50 μg |
| Type de cellule | Cancer Cells and Cell Lines; Neural Cells, PSC-Derived; Neural Stem and Progenitor Cells; Neurons |
| Pureté ou qualité | ≥95% |