Learn More
STEMCELL Technologies Human Recombinant Heregulin-beta 1
Code produit. 17139501
Encompass_Preferred
Click to view available options
Quantité:
10 μg
1 000 μg
50 μg
Code produit. 17139501
Marque: STEMCELL Technologies 78071.1
Heregulin-beta 1
  • Heregulin-beta 1 also known as neuregulin-1 (NRG-1) is a member of the epidermal growth factor (EGF) family of growth factors and acts as a ligand for ErbB family receptor tyrosine kinases
  • Heregulin/neuregulin is a family of structurally related polypeptide growth factors derived from alternatively spliced genes (NRG1, NRG2, NRG3, and NRG4)
  • Heregulin-beta 1 plays an important role during the development of the nervous system, heart, and mammary glands
  • Heregulin-beta 1 is expressed in neuronal cells, and modulates cell growth and differentiation of the cells during development and wound healing
  • It has been implicated through in vivo and in vitro studies that heregulin-beta 1/ErbB signaling is crucial for multiple aspects of cardiovascular development and protects the heart from ischemic injury
  • Heregulin-beta 1 also promotes invasiveness and metastasis of breast cancer cells
Spécification
Nom Acetylcholine receptor-inducing activity, ARIA, Breast cancer cell differentiation factor p45, Glial growth factor, Heregulin Neu differentiation factor, HRG, HRG1, HRG1-beta 1, Neuregulin-1, NRG1, NRG1-beta 1, Sensory and motor neuron-derived factor
Quantité 50 μg
Type de cellule Cancer Cells and Cell Lines; Neural Cells, PSC-Derived; Neural Stem and Progenitor Cells; Neurons
Pureté ou qualité ≥95%
Correction du contenu d'un produit
Le programme Encompass de Fisher Scientific propose des articles qui ne font pas partie de notre portefeuille de distribution. Ces produits n'ont généralement pas de photos ou de descriptions détaillées. Cependant, nous nous engageons à améliorer votre expérience d'achat. Veuillez utiliser le formulaire ci-dessous pour nous faire part de vos commentaires sur le contenu de ce produit.