Learn More
STEMCELL Technologies Human Recombinant IL-12
Click to view available options
Quantité:
1 000 μg
25 μg
5 μg
Conditionnement:
1000µg
25µg
5µg
Description
- Interleukin 12 (IL-12p70) is a heterodimeric cytokine composed of p35 and p40 subunits
- IL-12 is produced by monocytes, macrophages, dendritic cells, neutrophils, and B cells in response to bacterial products and cytokines such as IFN-γ
- The IL-12 receptor is expressed on T, NK, and dendritic cells
- Upon binding, IL-12 initiates signaling via the JAK/STAT signaling pathway and stimulates NK, B, and T cells to produce IFN-γ (Watford et al.) It also regulates cytokine synthesis, proliferation of T and NK cells, and stimulates differentiation of CD4+ and CD8+ T cells (Germann & Rüde)
- Mice that are deficient in IL-12 are susceptible to many intracellular pathogens and have impaired IFN-γ secretion, Th1 differentiation, and NK cytolytic activity; however, Th2 development and IL-4 production are enhanced (Watford et al.)
Spécification
Spécification
| Nom | CTL maturation factor, Cytotoxic lymphocyte maturation factor, IL-12p70, Interleukin-12, Natural killer cell stimulatory factor, NKSF |
| Quantité | 5 μg |
| Type de cellule | B Cells; Hematopoietic Stem and Progenitor Cells; Lymphocytes; NK Cells; T Cells |
| Pureté ou qualité | ≥95% |
Correction du contenu d'un produit
Le programme Encompass de Fisher Scientific propose des articles qui ne font pas partie de notre portefeuille de distribution. Ces produits n'ont généralement pas de photos ou de descriptions détaillées. Cependant, nous nous engageons à améliorer votre expérience d'achat. Veuillez utiliser le formulaire ci-dessous pour nous faire part de vos commentaires sur le contenu de ce produit.