Learn More
STEMCELL Technologies Human Recombinant IP-10 (CXCL10)
Code produit. p-7217411
Encompass_Preferred
Click to view available options
Quantité:
50 μg
Code produit. 17179731
Marque: STEMCELL Technologies 78165.1
Interferon gamma-inducible protein 10
  • Interferon gamma-inducible protein (IP) 10 or CXCL10 is a member of the CXC chemokine family
  • It binds CXCR3 activating ERK1/2, p38/MAPK, JNK, and PI3-kinase/AKT signaling pathways, inducing intracellular calcium influx, DNA synthesis, cell proliferation, and chemotaxis
  • IP-10 regulates innate and adaptive immune responses by affecting the function of activated T cells, natural killer cells, inflammatory dendritic cells, macrophages, and B cells
  • IP-10 is produced by leukocytes, activated neutrophils, eosinophils, monocytes, epithelial cells, endothelial cells, fibroblasts, and keratinocytes in response to IFN-gamma
  • IP-10 has been implicated in a wide range of diseases, including infectious diseases and chronic inflammatory and autoimmune diseases, as well as in tumor formation (Liu et al.)
Spécification
Nom 10 kDa interferon-gamma-induced protein, C7, crg-2, Chemokine (C-X-C motif) ligand 10, CXCL10, Gamma-IP-10, gIP-10, IFI10, INP10, mob1, SCYB10, Small inducible cytokine B10
Quantité 50 μg
Type de cellule Monocytes; T Cells
Pureté ou qualité ≥95%