Learn More
STEMCELL Technologies Human Recombinant RANTES (CCL5)
Click to view available options
Quantité:
100 μg
20 μg
Conditionnement:
100µg
20µg
Description
- RANTES (regulated upon activation, normal T cell expressed and secreted), also known as CCL5, is a member of the CC family of chemokines and is able to recruit leukocytes to sites of inflammation
- RANTES is secreted by T lymphocytes, macrophages, platelets, synovial fibroblasts, tubular epithelium, and certain types of tumor cells
- This chemokine exerts its effect by interacting with the chemokine receptors CCR1, CCR3, CCR4, and CCR5
- RANTES plays an active role in recruiting a variety of leukocytes into inflammatory sites, including T cells, macrophages, eosinophils, and basophils
- In collaboration with certain cytokines that are released by T cells such as IL-2 and IFN-γ, RANTES also induces the activation and proliferation of NK cells to generate CC chemokine-activated killer cells, which are highly cytolytic
Spécification
Spécification
| Nom | D17S136, Eosinophil chemotactic cytokine, Regulated upon activation normally T-expressed and secreted, SIS-delta, Small-inducible cytokine A5, T-cell specific protein p288 |
| Quantité | 100 μg |
| Type de cellule | B Cells; Dendritic Cells; Mesenchymal Stem and Progenitor Cells; Monocytes; NK Cells; Other; T Cells; T Cells, CD4+; T Cells, CD8+ |
| Pureté ou qualité | ≥95% |