Learn More
STEMCELL Technologies Human Recombinant RANTES (CCL5)
Code produit. 17189561
Encompass_Preferred
Click to view available options
Quantité:
100 μg
20 μg
Conditionnement:
100µg
20µg
Code produit. 17189561
Marque: STEMCELL Technologies 78099.1
Regulated upon activation, normal T cell expressed and secreted
  • RANTES (regulated upon activation, normal T cell expressed and secreted), also known as CCL5, is a member of the CC family of chemokines and is able to recruit leukocytes to sites of inflammation
  • RANTES is secreted by T lymphocytes, macrophages, platelets, synovial fibroblasts, tubular epithelium, and certain types of tumor cells
  • This chemokine exerts its effect by interacting with the chemokine receptors CCR1, CCR3, CCR4, and CCR5
  • RANTES plays an active role in recruiting a variety of leukocytes into inflammatory sites, including T cells, macrophages, eosinophils, and basophils
  • In collaboration with certain cytokines that are released by T cells such as IL-2 and IFN-γ, RANTES also induces the activation and proliferation of NK cells to generate CC chemokine-activated killer cells, which are highly cytolytic
Spécification
Nom D17S136, Eosinophil chemotactic cytokine, Regulated upon activation normally T-expressed and secreted, SIS-delta, Small-inducible cytokine A5, T-cell specific protein p288
Quantité 100 μg
Type de cellule B Cells; Dendritic Cells; Mesenchymal Stem and Progenitor Cells; Monocytes; NK Cells; Other; T Cells; T Cells, CD4+; T Cells, CD8+
Pureté ou qualité ≥95%
Correction du contenu d'un produit
Le programme Encompass de Fisher Scientific propose des articles qui ne font pas partie de notre portefeuille de distribution. Ces produits n'ont généralement pas de photos ou de descriptions détaillées. Cependant, nous nous engageons à améliorer votre expérience d'achat. Veuillez utiliser le formulaire ci-dessous pour nous faire part de vos commentaires sur le contenu de ce produit.