Learn More
STEMCELL Technologies Human Recombinant Shh (C24II)
Click to view available options
Quantité:
10 μg
1 000 μg
50 μg
Conditionnement:
50µg
Description
- Sonic hedgehog (Shh) is a member of the Hedgehog family of secreted signaling proteins that has a prominent role in patterning during early mammalian development (Ho & Scott)
- It is expressed during early embryogenesis in a variety of tissues (Finco et al.) During patterning of the central nervous system, Shh directly acts on cells to specify neural cell fates, regulates proliferation and survival of oligodendrocytes precursors and neural crest cells and stimulates proliferation, differentiation and axon growth (Ho & Scott; Martí)
- Shh also regulates T cell differentiation and activation (Crompton et al.) Recombinant human Shh (C24II; rhShh) is a fully biologically active molecule
Spécification
Spécification
| Nom | HHG-1, HLP3, HPE3, MCOPCB5, SMMCI, Sonic hedgehog, TPT, TPTPS |
| Quantité | 50 μg |
| Type de cellule | Neural Cells, PSC-Derived; Neural Stem and Progenitor Cells; Pluripotent Stem Cells |
| Pureté ou qualité | ≥95% |