missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human Semaphorin 3A Fc Recombinant Protein, PeproTech®

Code produit. 17881663
Click to view available options
Quantité:
10 x 100 μg
100 μg
25 μg
5 μg
5 x 100 μg
Conditionnement:
100µg
1mg
25µg
5 pièces
500µg
5µg
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Code produit. 17881663

Marque: Gibco™ 15017H100UG

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Recombinant Protein

150-17H-1MG will be provided as 10 x 100 μg (150-17H-100UG). 150-17H-500UG will be provided as 5 x 100 μg (150-17H-100UG). Recombinant Human Semaphorin 3A Fc has a calculated molecular weight of 226.2 kDa and therefore runs above the 200kDa marker by SDS-PAGE analysis under nonreducing conditions. When run under reducing conditions, this protein migrates as three distinct bands that, due to glycosylation, run higher than expected at apparent molecular weights of approximately 120-130 kDa, 90-100 kDa, and 35-40 kDa. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

This gene is a member of the semaphorin family and encodes a protein with an Ig-like C2-type domain, a PSI domain and a Sema domain. This secreted protein can function as either a chemorepulsive agent, inhibiting axonal outgrowth, or as a chemoattractive agent, stimulating the growth of apical dendrites. In both cases, the protein is vital for normal neuronal pattern development. Increased expression of this protein is associated with schizophrenia and is seen in a variety of human tumor cell lines. Also, aberrant release of this protein is associated with the progression of Alzheimer's disease.
TRUSTED_SUSTAINABILITY

Spécification

Numéro d’adhésion Q14563
À utiliser avec (application) Control, ELISA Standard, Binding Assay
Formule Protein with no preservative
Identification génétique (Entrez) 10371
Poids moléculaire 226.2 kDa
Nom Human Semaphorin 3A Fc
Méthode de purification Purified
Quantité 100 μg
Source CHO cells
Conditions de stockage -20°C
État réglementaire RUO
Concentration en endotoxines <1 EU/μg
Alias de gène COLL1; coll-1; collapsin 1; collapsin-1; HH16; Hsema-I; Hsema-III; MGC133243; sema D; sema domain, immunoglobulin domain (Ig), short basic domain, sec; sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3A; sema III; SEMA1; SEMA3A; Semad; SEMAIII; SEMAL; Semaphorin 3a; semaphorin D; semaphorin III; semaphorin L; semaphorin-3A; Semaphorin-D; SemD
Nom usuel SEMA3A
Symbole de gène(s) SEMA3A
Activité biologique Determined by its ability to bind recombinant rat Neuropilin-1 Fc Chimera in a functional ELISA assay.
Type de produit Protein
Conjugué Unconjugated
Recombinant Recombinant
Marqueur de protéine Fc-tag, His-tag
Séquence HHHHHHHHGK NNVPRLKLSY KEMLESNNVI TFNGLANSSS YHTFLLDEER SRLYVGAKDH IFSFDLVNIK DFQKIVWPVS YTRRDECKWA GKDILKECAN FIKVLKAYNQ THLYACGTGA FHPICTYIEI GHHPEDNIFK LENSHFENGR GKSPYDPKLL TASLLIDGEL YSGTAADFMG RDFAIFRTLG HHHPIRTEQH DSRWLNDPKF ISAHLISESD NPEDDKVYFF FRENAIDGEH SGKATHARIG QICKNDFGGH RSLVNKWTTF LKARLICSVP GPNGIDTHFD ELQDVFLMNF KDPKNPVVYG VFTTSSNIFK GSAVCMYSMS DVRRVFLGPY AHRDGPNYQW VPYQGRVPYP RPGTCPSKTF GGFDSTKDLP DDVITFARSH PAMYNPVFPM NNRPIVIKTD VNYQFTQIVV DRVDAEDGQY DVMFIGTDVG TVLKVVSIPK ETWYDLEEVL LEEMTVFREP TAISAMELST KQQQLYIGST AGVAQLPLHR CDIYGKACAE CCLARDPYCA WDGSACSRYF PTAKRATRAQ DIRNGDPLTH CSDLHHDNHH GHSPEERIIY GVENSSTFLE CSPKSQRALV YWQFQRRNEE RKEEIRVDDH IIRTDQGLLL RSLQQKDSGN YLCHAVEHGF IQTLLKVTLE VIDTEHLEEL LHKDDDGDGS KTKEMSNSMT PSQKVWYRDF MQLINHPNLN TMDEFCEQVW KRDRKQRRQR PGHTPGNSNK WKHLQENKKG RNRRTHEFER APRSVGGPKS CDKTHTCPPC PAPELLGGPS VFLFPPKPKD TLMISRTPEV TCVVVDVSHE DPEVKFNWYV DGVEVHNAKT KPREEQYNST YRVVSVLTVL HQDWLNGKEY KCKVSNKALP APIEKTISKA KGQPREPQVY TLPPSRDELT KNQVSLTCLV KGFYPSDIAV EWESNGQPEN NYKTTPPVLD SDGSFFLYSK LTVDKSRWQQ GNVFSCSVMH EALHNHYTQK SLSLSPGK
Contenu et stockage -20°C
Activité Determined by its ability to bind recombinant rat Neuropilin-1 Fc Chimera in a functional ELISA assay.
Système d’expression CHO cells
Forme Lyophilized
Pureté ou qualité ≥ 95% by SDS-PAGE gel and HPLC analyses.
Afficher plus Afficher moins
Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis