missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human SMARCA1 (aa 500-601) Control Fragment Recombinant Protein
Click to view available options
Quantité:
100 μl
Conditionnement:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (97%), Rat (97%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-51899 (PA5-51899. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Members of this G protein family are characterized by the presence of conserved SNF2 domain, a helicase-like region. They also have an ATPase activity and are thought to have a significant role regulating chromatin remodeling. Although SNF2L1 has a ubiquitous expression pattern, its active form is expressed in neurons suggesting a possible role in promoting neurite outgrowth and regulating neuronal physiology. Principal Names: SWI/SNF related matrix associated actin dependent regulator of chromatin subfamily A member 1 (SMARCA1); SNF2L1; SNF2LB; Possible global transcription activator SNF2L1; SWI2 Official Gene Symbol: SMARCA1 Gen Bank Accession Number: NP_620604 Gene ID: 6594 (Human) Gene Map Locus: Xq25 (Human) SNF2L1, a ∽123KDa protein, is a member of SWI/SNF family of proteins.
Spécification
Spécification
| Numéro d’adhésion | P28370 |
| Concentration | ≥5.0 mg/mL |
| À utiliser avec (application) | Blocking Assay, Control |
| Formule | 1 M urea, PBS with no preservative; pH 7.4 |
| Identification génétique (Entrez) | 6594 |
| Nom | Human SMARCA1 (aa 500-601) Control Fragment |
| Quantité | 100 μl |
| État réglementaire | RUO |
| Alias de gène | 5730494M04Rik; ATP-dependent helicase SMARCA1; DKFZp686D1623; DNA-dependent ATPase SNF2L; FLJ41547; global transcription activator homologous sequence; hSNF2L; ISWI; nucleosome-remodeling factor subunit SNF2L; NURF140; Probable global transcription activator SNF2L1; RGD1561046; RP3-353H6.2; SMARCA1; SNF2L; SNF2L1; SNF2LB; SNF2-like 1; SNF2L-like protein; SNF2LT; sucrose nonfermenting 2-like protein 1; SWI; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 1; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 1; SWI2 |
| Nom usuel | SMARCA1 |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu