Learn More
Novus Biologicals™ Human Stomach Whole Tissue Lysate (Adult Whole Primary and Metastatic Tumor Paired)
Code produit 18268970 Tous les produits Bio Techne Produits
Modifier l'affichage
Click to view available options
Quantité:
1 jeu
Conditionnement:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
18268970 1 jeu 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 18268970 Fournisseur Novus Biologicals™ Code fournisseur NBL126533
Please to purchase this item. Need a web account? Register with us today!
The Human Stomach Whole Tissue Lysate (Adult Whole Primary and Metastatic Tumor Paired) is derived from human.
TRUSTED_SUSTAINABILITY
Spécification
Tissue Digestive/Waste
Quantité 1 jeu
Tampon de stockage HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors
Spécificité du test Matched Pair - Total Protein - Human Primary Tumor and Metastatic Tumor Tissue (PM): Stomach
Fraction cellulaire Whole
État des tissus Primary and Metastatic Tumor Paired
Réactivité croisée Human
Espèces hôtes Human
Type de produit Human Stomach Whole Tissue Lysate (Adult Primary and Metastatic Tumor Paired)
Contenu et stockage -80°C. Avoid freeze-thaw cycles.
Étape du cycle de vie Adulte
Protein State Native
Afficher plus Afficher moins
Usage exclusivement réservé à la recherche