missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human TNF-alpha, Animal-Free Recombinant Protein, PeproTech®
Click to view available options
Quantité:
1 mg
10 μg
100 μg
500 μg
Conditionnement:
100µg
10µg
1mg
500µg
Description
Recombinant Human TNF-alpha is a soluble 157 amino acid protein (17.4 kDa) which corresponds to C-terminal extracellular domain of the full length transmembrane protein. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
TNF alpha is a multifunctional proinflammatory cytokine that belongs to the tumor necrosis factor (TNF) superfamily. This cytokine is mainly secreted by macrophage and bind to its receptors, TNFRSF1A/TNFR1 and TNFRSF1B/TNFBR. TNF alpha is involved in the regulation of immune cells, cell proliferation, differentiation, apoptosis, lipid metabolism, and coagulation. TNF alpha exists as a multimer of two, three, or five noncovalently linked units, but shows a single 17 kDa band following SDS-PAGE under non-reducing conditions. Knockout studies in mice also suggested the neuroprotective function of TNF alpha, and has been observed to causes tumor necrosis when injected into tumor-bearing mice. Other functions of TNF-alpha include its role in the immune response to bacterial, viral, parasitic and certain fungal infections, as well as its role in the necrosis of specific tumors. TNF alpha causes cytolysis or cytostasis of certain transformed cells, being synergistic with interferon-gamma in its cytotoxicity. This cytokine has been implicated in a variety of diseases, including autoimmune diseases, insulin resistance, and cancer.
Spécification
Spécification
| Numéro d’adhésion | P01375 |
| À utiliser avec (application) | Functional Assay |
| Formule | protein with no preservative |
| Identification génétique (Entrez) | 7124 |
| Poids moléculaire | 17.4 kDa |
| Nom | Human TNF-alpha, Animal-Free |
| Quantité | 100 μg |
| Source | E. coli |
| État réglementaire | RUO |
| Concentration en endotoxines | <0.1 EU/ μg |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu