missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human TTF2 (aa 58-145) Control Fragment Recombinant Protein
Click to view available options
Quantité:
100 μl
Conditionnement:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (61%), Rat (61%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-84850 (PA5-84850. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
This gene encodes a member of the SWI2/SNF2 family of proteins, which play a critical role in altering protein-DNA interactions. The encoded protein has been shown to have dsDNA-dependent ATPase activity and RNA polymerase II termination activity. This protein interacts with cell division cycle 5-like, associates with human splicing complexes, and plays a role in pre-mRNA splicing.
Spécification
Spécification
| Numéro d’adhésion | Q9UNY4 |
| Concentration | ≥5.0 mg/mL |
| À utiliser avec (application) | Blocking Assay, Control |
| Formule | 1 M urea, PBS with no preservative; pH 7.4 |
| Identification génétique (Entrez) | 8458 |
| Nom | Human TTF2 (aa 58-145) Control Fragment |
| Quantité | 100 μl |
| État réglementaire | RUO |
| Alias de gène | 4632434F22Rik; 8030447N19; AV218430; F2; HuF2; human factor 2; lodestar homolog; lodestar protein; RNA polymerase II termination factor; Transcription release factor 2; transcription termination factor 2; transcription termination factor, RNA polymerase II; Ttf2; ZGRF6; zinc finger, GRF-type containing 6 |
| Nom usuel | TTF2 |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu