missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human TWEAK, Animal-Free Recombinant Protein, PeproTech®

Code produit. 17803573
Click to view available options
Quantité:
1 mg
100 μg
25 μg
500 μg
Conditionnement:
100µg
1mg
25µg
500µg
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Code produit. 17803573

Marque: Gibco™ AF310061MG

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Recombinant Protein

Recombinant Human TWEAK is a soluble 17.0 kDa polypeptide (154 amino acid residues) comprising the TNF-homologous region of TWEAK, and is generated by proteolytic processing of the full length membrane-anchored TWEAK protein. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

TNF-related weak inducer of apoptosis (TWEAK) is a member of the tumor necrosis factor superfamily (TNFSF) of structurally related cytokines. Like most other members of this family, TWEAK is a cell surface-associated type II transmembrane protein although a smaller, biologically active form can also be generated by cleavage near the cell membrane. TWEAK has multiple biological activities, including stimulation of cell growth and angiogenesis, induction of inflammatory cytokines, in addition to stimulation of apoptosis. The TWEAK signal transduction pathway has not been well established but it appears to signal via TweakR/Fn14 in a manner similar to that described for other TNFSF members that bind receptors lacking death domains.
TRUSTED_SUSTAINABILITY

Spécification

Numéro d’adhésion O43508
À utiliser avec (application) Functional Assay
Formule protein with no preservative
Identification génétique (Entrez) 8742
Poids moléculaire 17 kDa
Nom Human TWEAK, Animal-Free
Quantité 1 mg
Source E. coli
État réglementaire RUO
Concentration en endotoxines <0.1 EU/ μg
Alias de gène APO3 ligand; APO3/DR3 ligand; APO3L; Dr3l; Dr3lg; LOW QUALITY PROTEIN: tumor necrosis factor ligand superfamily member 12; MGC129581; MGC20669; TNF superfamily member 12; TNF-related weak inducer of apoptosis; TNFSF12; TNLG4A; tumor necrosis factor (ligand) superfamily member 12; tumor necrosis factor (ligand) superfamily, member 12; tumor necrosis factor ligand 4 A; tumor necrosis factor ligand superfamily member 12; Tumor necrosis factor ligand superfamily member 12, membrane form; Tumor necrosis factor ligand superfamily member 12, secreted form; tumor necrosis factor superfamily member 12; TWEAK; UNQ181/PRO207; zgc:153941
Nom usuel TWEAK
Symbole de gène(s) Tnfsf12
Activité biologique Assay 1: The ED50 as determined by the dose-dependent stimulation of IL-8 production by human PBMC is less than 10 ng/ml Assay 2: TWEAK weakly induces the death of HT29 cells when cultured in the presence of IFN-gamma. The ED50 for this effect is between 30-45 ng/ml
Conjugué Unconjugated
Recombinant Recombinant
Séquence MKGRKTRARR AIAAHYEVHP RPGQDGAQAG VDGTVSGWEE ARINSSSPLR YNRQIGEFIV TRAGLYYLYC QVHFDEGKAV YLKLDLLVDG VLALRCLEEF SATAASSLGP QLRLCQVSGL LALRPGSSLR IRTLPWAHLK AAPFLTYFGL FQVH
Contenu et stockage -20°C
Système d’expression E. coli
Forme Lyophilized
Pureté ou qualité ≥ 98% by SDS-PAGE gel and HPLC analyses.
Afficher plus Afficher moins
Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis