missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human VEGF-D Recombinant Protein, PeproTech®
Click to view available options
Quantité:
1 mg
10 μg
100 μg
2 μg
250 μg
500 μg
Conditionnement:
100µg
10µg
1mg
250µg
2µg
500µg
Description
Recombinant Human VEGF-D is a 26.2 kDa, non-disulfide linked, homodimeric protein consisting of two 117 amino acid polypeptide chains. Due to glycosylation, the protein migrates as a 20.0-22.0 kDa band by SDS-PAGE analysis under non-reducing conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
The protein encoded by this gene is a member of the platelet-derived growth factor/vascular endothelial growth factor (PDGF/VEGF) family and is active in angiogenesis, lymphangiogenesis, and endothelial cell growth. This secreted protein undergoes a complex proteolytic maturation, generating multiple processed forms which bind and activate VEGFR-2 and VEGFR-3 receptors. This protein is structurally and functionally similar to vascular endothelial growth factor C.
Spécification
Spécification
| Numéro d’adhésion | O43915 |
| À utiliser avec (application) | Control, ELISA Standard, Binding Assay, Bioactivity |
| Formule | Protein with no preservative |
| Identification génétique (Entrez) | 2277 |
| Poids moléculaire | 26.2 kDa |
| Nom | Human VEGF-D |
| Méthode de purification | Purified |
| Quantité | 100 μg |
| Source | HEK293 cells |
| Conditions de stockage | -20°C |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu