missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human VWA5A (aa 155-248) Control Fragment Recombinant Protein
Click to view available options
Quantité:
100 μl
Conditionnement:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (74%), Rat (74%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
May play a role in tumorigenesis as a tumor suppressor. Altered expression of this protein and disruption of the molecular pathway it is involved in, may contribute directly to or modify tumorigenesis.
Spécification
Spécification
| Numéro d’adhésion | O00534 |
| Concentration | 3.90 mg/mL |
| À utiliser avec (application) | Neutralization, Control |
| Formule | PBS, 1M urea with no preservative; pH 7.4 |
| Identification génétique (Entrez) | 4013 |
| Nom | Human VWA5A (aa 155-248) Control Fragment |
| Plage de pH | 7.4 |
| Méthode de purification | Metal-chelate chromatography |
| Quantité | 100 μl |
| Conditions de stockage | -20°C, Avoid Freeze/Thaw Cycles |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu